2,3-Diaminotoluene, 97%
2,3-Diaminotoluene, 97%
2,3-Diaminotoluene, 97%
2,3-Diaminotoluene, 97%
Thermo Scientific Chemicals

2,3-Diaminotoluene, 97%

CAS: 2687-25-4 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15647.06
also known as A15647-06
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.30
Save 13.65 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.30
Save 13.65 (15%)
Each
Add to cart
Chemical Identifiers
CAS2687-25-4
IUPAC Name3-methylbenzene-1,2-diamine
Molecular FormulaC7H10N2
InChI KeyAXNUJYHFQHQZBE-UHFFFAOYSA-N
SMILESCC1=CC=CC(N)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to brown to black
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)54.0-63.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only