2,6-Diaminopurine, 98%
2,6-Diaminopurine, 98%
2,6-Diaminopurine, 98%
Thermo Scientific Chemicals

2,6-Diaminopurine, 98%

CAS: 1904-98-9 | C5H6N6 | 150.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A15649.06
also known as A15649-06
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1904-98-9
IUPAC Name7H-purine-2,6-diamine; sulfuric acid
Molecular FormulaC5H8N6O4S
InChI KeyKYYLASPNTMEZTB-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC(N)=C2NC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only