Thermo Scientific Chemicals

1,8-Dibromooctane, 98%, Thermo Scientific Chemicals

Catalog number: A15651.18
50 g, Each
Thermo Scientific Chemicals

1,8-Dibromooctane, 98%, Thermo Scientific Chemicals

Catalog number: A15651.18
50 g, Each
Quantity
Catalog number: A15651.18
also known as A15651-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4549-32-0
IUPAC Name
1,8-dibromooctane
Molecular Formula
C8H16Br2
InChI Key
DKEGCUDAFWNSSO-UHFFFAOYSA-N
SMILES
BrCCCCCCCCBr
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.4975-1.5015 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text