1-Hexylamine, 99%
1-Hexylamine, 99%
1-Hexylamine, 99%
Thermo Scientific Chemicals

1-Hexylamine, 99%

CAS: 111-26-2 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
Catalog number A15663.AU
also known as A15663-AU
Price (USD)
260.00
Each
Quantity:
1000 mL
Request bulk or custom format
Price (USD)
260.00
Each
Chemical Identifiers
CAS111-26-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4165-1.4195 @ 20°C
Identification (FTIR)Conforms (non-U.S. specification)
FormLiquid
View more
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

Solubility
Miscible with water, dichloromethane, alcohol and acetone.

Notes
Air sensitive. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kizaki, K.; Imoto, H.; Kato, T.; Naka, K. Facile construction of N-alkyl arylaminomaleimide derivatives as intensively emissive aggregation induced emission dyes. Tetrahedron 2015, 71 (4), 643-647.
  2. Navia, P.; Bessieres, D.; Plantier, F. Heat capacity of 1-pentylamine and 1-hexylamine: Experimental determination and modeling through a two-state association model (TSAM). J. Chem. Thermodyn. 2013, 57, 367-371.