4-Benzyl-4-hydroxypiperidine, 97%
4-Benzyl-4-hydroxypiperidine, 97%
4-Benzyl-4-hydroxypiperidine, 97%
4-Benzyl-4-hydroxypiperidine, 97%
Thermo Scientific Chemicals

4-Benzyl-4-hydroxypiperidine, 97%

CAS: 51135-96-7 | C12H17NO | 191.274 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15664.03
also known as A15664-03
Price (USD)
23.65
Online Exclusive
26.60
Save 2.95 (11%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
23.65
Online Exclusive
26.60
Save 2.95 (11%)
Each
Add to cart
Chemical Identifiers
CAS51135-96-7
IUPAC Name4-benzyl-4-hydroxypiperidin-1-ium
Molecular FormulaC12H18NO
InChI KeyKJZBZOFESQSBCV-UHFFFAOYSA-O
SMILESOC1(CC2=CC=CC=C2)CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤2.5%
Assay (GC)≥96.0%
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Laura C. Cooper, et al. 2-Aryl Indole NK1 receptor antagonists: optimisation of indole substitution.Bioorg. Med. Chem. Lett.,2009,11(9), 1233-1236.
  2. Duncan Shawa, et al. 2-Aryl indole NK1 antagonists: optimisation of the amide substituent.Bioorg. Med. Chem. Lett.,2001,11(23), 3031-3034.