3-Phenoxybenzyl chloride, 97%
3-Phenoxybenzyl chloride, 97%
3-Phenoxybenzyl chloride, 97%
Thermo Scientific Chemicals

3-Phenoxybenzyl chloride, 97%

CAS: 53874-66-1 | C13H11ClO | 218.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15670.06
also known as A15670-06
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS53874-66-1
IUPAC Name1-(chloromethyl)-3-phenoxybenzene
Molecular FormulaC13H11ClO
InChI KeyQUYVTGFWFHQVRO-UHFFFAOYSA-N
SMILESClCC1=CC=CC(OC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5880-1.5930 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only