Methyl anthranilate, 99%
Thermo Scientific Chemicals

Methyl anthranilate, 99%

CAS: 134-20-3 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15681.0B
also known as A15681-0B
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
106.00
Save 15.35
Each
Chemical Identifiers
CAS134-20-3
IUPAC Namemethyl 2-aminobenzoate
Molecular FormulaC8H9NO2
InChI KeyVAMXMNNIEUEQDV-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.5815-1.5845 @ 20°C (non-U.S. specification)
View more
It is applied as a flavoring agent. Food companies use artificial MA in their candy and chewing gum to give them their grape flavor. Beverage companies have been adding it to the carbonated water and sugar in their soft drinks for the same reason.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.