It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only
General References:
- A. Segal, et al. Formation of 6-dimethylcarbamyloxy-dGuo, 6-dimethylamino-dGuo and 4-dimethylamino-dThd following in vitro reaction of dimethylcarbamyl chloride with calf thymus DNA and 6-diethylcarbamyloxy-dGuo following in vitro reaction of diethylcarbamyl chloride with calf thymus DNA.Chem. Biol. Interact.,1982,40(2), 209-231.
- S. Kushner, et al. Experimental chemotherapy of filariasis. v. the preparation of derivatives of piperazine.J. Org. Chem.,1948,13(1), 144-153.