9,10-Diphenylanthracene, 99%
9,10-Diphenylanthracene, 99%
9,10-Diphenylanthracene, 99%
Thermo Scientific Chemicals

9,10-Diphenylanthracene, 99%

CAS: 1499-10-1 | C26H18 | 330.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15714.14
also known as A15714-14
Price (USD)
661.65
Special offer
Online exclusive
Ends: 30-Jun-2026
778.00
Save 116.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
661.65
Special offer
Online exclusive
Ends: 30-Jun-2026
778.00
Save 116.35 (15%)
Each
Chemical Identifiers
CAS1499-10-1
IUPAC Name9,10-diphenylanthracene
Molecular FormulaC26H18
InChI KeyFCNCGHJSNVOIKE-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Assay (HPLC)>98.5%
Appearance (Color)Yellow to yellow-green
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Lee, S. B.; Park, S. N.; Kim, C.; Lee, H. W.; Lee, H. W.; Kim, Y. K.; Yoon, S. S. Synthesis and electroluminescent properties of 9,10-diphenylanthracene containing 9H-carbazole derivatives for blue organic light-emitting diodes. Synth. Met. 2015, 203, 174-179.
  2. Berger, R.; Wagner, M.; Feng, X.; Müllen, K. Polycyclic aromatic azomethine ylides: a unique entry to extended polycyclic heteroaromatics. Chem. Sci. 2015, 6, 436-441.