4-Benzyloxybenzoic acid, 98%
4-Benzyloxybenzoic acid, 98%
4-Benzyloxybenzoic acid, 98%
Thermo Scientific Chemicals

4-Benzyloxybenzoic acid, 98%

CAS: 1486-51-7 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15715.03
also known as A15715-03
Price (USD)
22.65
Online Exclusive
24.70
Save 2.05 (8%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
22.65
Online Exclusive
24.70
Save 2.05 (8%)
Each
Chemical Identifiers
CAS1486-51-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (clear melt)186.0-194.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only