2-Amino-5-chlorobenzophenone, 98+%
2-Amino-5-chlorobenzophenone, 98+%
2-Amino-5-chlorobenzophenone, 98+%
Thermo Scientific Chemicals

2-Amino-5-chlorobenzophenone, 98+%

CAS: 719-59-5 | C13H10ClNO | 231.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A15720.22
also known as A15720-22
Price (USD)
43.65
Online Exclusive
48.10
Save 4.45 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.10
Save 4.45 (9%)
Each
Chemical Identifiers
CAS719-59-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%
Melting Point (clear melt)95.0-102.0?C
Identification (FTIR)Conforms
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. R. Ramesh Babu, et al. Studies on organic single crystal: 2-amino-5-chlorobenzophenone (2A-5CB).J. Cryst. Growth.,2004,256(1-2), 290-295.
  2. M. Gulam Mohamed, et al. Growth and characterization of 2-amino-5-chlorobenzophenone (2A-5CB) single crystals.J. Cryst. Growth.,2007,300(2), 409-414.