n-Pentylbenzene, 96%
n-Pentylbenzene, 96%
n-Pentylbenzene, 96%
n-Pentylbenzene, 96%
Thermo Scientific Chemicals

n-Pentylbenzene, 96%

CAS: 538-68-1 | C11H16 | 148.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15734.14
also known as A15734-14
Price (USD)
72.65
Online Exclusive
80.60
Save 7.95 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.60
Save 7.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS538-68-1
IUPAC Namepentylbenzene
Molecular FormulaC11H16
InChI KeyPWATWSYOIIXYMA-UHFFFAOYSA-N
SMILESCCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4850-1.4910 @ 20?C
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only