Ethylene carbonate, 99%
Ethylene carbonate, 99%
Ethylene carbonate, 99%
Ethylene carbonate, 99%
Thermo Scientific™

Ethylene carbonate, 99%

CAS: 96-49-1 | C3H4O3 | 88.062 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A15735.0C
also known as A15735-0C
Price (USD)
336.65
Special offer
Online exclusive
Ends: 30-Jun-2026
396.00
Save 59.35 (15%)
Each
Quantity:
10,000 g
Request bulk or custom format
Price (USD)
336.65
Special offer
Online exclusive
Ends: 30-Jun-2026
396.00
Save 59.35 (15%)
Each
Chemical Identifiers
CAS96-49-1
IUPAC Name1,3-dioxolan-2-one
Molecular FormulaC3H4O3
InChI KeyKMTRUDSVKNLOMY-UHFFFAOYSA-N
SMILESO=C1OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
CommentMaterial Sourced in UK and US
Melting Point (clear melt)34.0-40.0?C
Appearance (Color)Colorless to white to yellow
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
View more
Solvent for many polymers and resins, plasticisers, rubbers and textiles.Ethylene carbonate is used as a polar solvent and a high permittivity component of electrolytes in lithium batteries. It finds application in surface coatings, dyes, fibers and plastics. It is also used as a plasticizer and as a precursor to vinylene carbonate. Further, it is used in the preparation of dimethyl carbonate, which acts as a useful solvent and a mild methylating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent for many polymers and resins, plasticisers, rubbers and textiles.Ethylene carbonate is used as a polar solvent and a high permittivity component of electrolytes in lithium batteries. It finds application in surface coatings, dyes, fibers and plastics. It is also used as a plasticizer and as a precursor to vinylene carbonate. Further, it is used in the preparation of dimethyl carbonate, which acts as a useful solvent and a mild methylating agent.

Solubility
Miscible (40%) with water, alcohol ethyl acetate, benzene and chloroform. Soluble in ether, n-Butanol, and carbon tetrachloride.Soluble in water, n-butanol, ethyl acetate, benzene, carbon tetrachloride, ether and chloroform. Insoluble in gasoline and turpentine oil.

Notes
Incompatible with strong oxidizing agents, acids, bases and reducing agents. Store in a cool place.
RUO – Research Use Only

General References:

  1. Ethylene oxide equivalent for introduction of the 2-hydroxyethyl group, by reaction with various nucleophiles. Reacts with thiols (K2CO3/ 18-crown-6) to give 2-hydroxyethyl sulfides: Synthesis, 382 (1986).
  2. You, X.; Chaudhari, M.; Rempe, S.; Pratt, L. R. Dielectric Properties of Ethylene Carbonate and Propylene Carbonate Using Molecular Dynamics Simulations. ECS Trans. 2015, 69 (1), 107-111.
  3. Chen, X.; Cui, Y.; Wen, C.; Wang, B.; Dai, W. L. Continuous synthesis of methanol: heterogeneous hydrogenation of ethylene carbonate over Cu/HMS catalysts in a fixed bed reactor system. Chem. Commun. 2015, 51 (72), 13776-13778.