2-Thiophenemethanol, 97%
2-Thiophenemethanol, 97%
2-Thiophenemethanol, 97%
Thermo Scientific Chemicals

2-Thiophenemethanol, 97%

CAS: 636-72-6 | C5H6OS | 114.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A15768.14
also known as A15768-14
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
61.10
Save 9.45
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
61.10
Save 9.45
Each
Chemical Identifiers
CAS636-72-6
IUPAC Name(thiophen-2-yl)methanol
Molecular FormulaC5H6OS
InChI KeyZPHGMBGIFODUMF-UHFFFAOYSA-N
SMILESOCC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5615-1.5665 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only