Phenazine, 99+%
Phenazine, 99+%
Phenazine, 99+%
Thermo Scientific Chemicals

Phenazine, 99+%

CAS: 92-82-0 | C12H8N2 | 180.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15770.22
also known as A15770-22
Price (USD)
478.65
Online Exclusive
532.00
Save 53.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
478.65
Online Exclusive
532.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS92-82-0
IUPAC Namephenazine
Molecular FormulaC12H8N2
InChI KeyPCNDJXKNXGMECE-UHFFFAOYSA-N
SMILESC1=CC2=NC3=CC=CC=C3N=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
FormCrystals or powder or crystalline powder or needles
Identification (FTIR)Conforms
Melting Point (clear melt)171.0-177.0?C
Assay (GC)≥99.0%
Phenazine is the parent substance of many dyestuffs, such as the toluylene red, indulines and safranines. It can be applied in therapeutic uses, used as antipsychotic and antiemetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Morimitsu Nishikimi.; N. Appaji Rao.; Kunio Yagi. The occurrence of superoxide anion in the reaction of reduced phenazine methosulfate and molecular oxygen. Biochem. Biophys. Res. Commun. 1972, 46 (2), 849-854 .
  2. Korneel Rabaey.; Nico Boon.; Monica Höfte .; Willy Verstraete. Microbial Phenazine Production Enhances Electron Transfer in Biofuel Cells.Environ. Sci. Technol. 2005, 39 (9),3401-3408 .