CD147 Monoclonal Antibody (MEM-M6/1), PE - FAQs
CD147 Monoclonal Antibody (MEM-M6/1), PE - FAQs
View additional product information for CD147 Monoclonal Antibody (MEM-M6/1), PE - FAQs (A15772)
1 product FAQs found
Is the CD147 Monoclonal Antibody (MEM-M6/1), PE (Cat. No. A15772) a blocking antibody that hampers the biological function of the surface antigen when bound to it?
Sorry, we have not analyzed this.