4-Methoxybenzaldehyde dimethyl acetal, 98%
4-Methoxybenzaldehyde dimethyl acetal, 98%
4-Methoxybenzaldehyde dimethyl acetal, 98%
Thermo Scientific Chemicals

4-Methoxybenzaldehyde dimethyl acetal, 98%

CAS: 2186-92-7 | C10H14O3 | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
500 g
Catalog number A15793.30
also known as A15793-30
Price (USD)
296.65
Online Exclusive
329.00
Save 32.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
296.65
Online Exclusive
329.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS2186-92-7
IUPAC Name1-(dimethoxymethyl)-4-methoxybenzene
Molecular FormulaC10H14O3
InChI KeyNNHYAHOTXLASEA-UHFFFAOYSA-N
SMILESCOC(OC)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.5035-1.5075 @ 20°C
4-Methoxybenzaldehyde dimethyl acetal is used as a precursor to 1-methoxy-1-(4-methoxyphenyl)hept-2-yne, 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate and 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene. Further, it acts as a protecting group reagent for diols, especially in carbohydrates. It is also used as a flavor essence in sunflower, cyclamen and in jasmine. In addition to this, it is involved in allylation reactions with allyltrimethylsilane catalyzed by Iron(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzaldehyde dimethyl acetal is used as a precursor to 1-methoxy-1-(4-methoxyphenyl)hept-2-yne, 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate and 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene. Further, it acts as a protecting group reagent for diols, especially in carbohydrates. It is also used as a flavor essence in sunflower, cyclamen and in jasmine. In addition to this, it is involved in allylation reactions with allyltrimethylsilane catalyzed by Iron(III) chloride.

Solubility
Miscible with alcohol and many organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
RUO – Research Use Only

General References:

  1. Reagent for the protection of diols as their cyclic acetals, hydrolyzed ca 10x faster by acid than the benzylidene acetal: Rec. Trav. Chim., 103, 335 (1984); J. Chem. Soc., Perkin 1, 2371 (1984).
  2. Li, Q. F.; Lin, H. X.; Cui, Y. M.; Xu, P. P. Syntheses and biological evaluation of C-3'-N-acyl modified taxane analogues from 1-deoxybaccatin-VI. Eur. J. Med. Chem. 2015, 104, 97-105.
  3. Li, P.; Yu, Y.; Huang, P. P.; Liu, H.; Cao, C. Y.; Song, W. G. Core-shell structured MgAl-LDO@Al-MS hexagonal nanocomposite: an all inorganic acid-base bifunctional nanoreactor for one-pot cascade reactions. J. Mater. Chem. A 2014, 2 (2), 339-344.