Quercetin dihydrate, 97%
Quercetin dihydrate, 97%
Quercetin dihydrate, 97%
Thermo Scientific Chemicals

Quercetin dihydrate, 97%

CAS: 6151-25-3 | C15H14O9 | 338.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15807.36
also known as A15807-36
Price (USD)
485.65
Special offer
Online exclusive
Ends: 30-Jun-2026
571.00
Save 85.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
485.65
Special offer
Online exclusive
Ends: 30-Jun-2026
571.00
Save 85.35 (15%)
Each
Chemical Identifiers
CAS6151-25-3
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Water Content (Karl Fischer Titration)10.1 - 11.8%
Melting Point>300?C
Appearance (Color)Bright yellow
View more
Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.

Solubility
Soluble in DMSO (68 mg/ml at 25°C), Acetic Acid (1 mg/ml), Aqueous Alkali (1 mg/ml), water (<1 at 25°C), and ethanol (21 mg/ml at 25°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Song-Hae Bok; Sun-Young Park; Yong Bok Park; Mi-Kyung Lee; Seon-Min Jeon; Tae-Sook Jeong; Myung-Sook Choi. Quercetin Dihydrate and Gallate Supplements Lower Plasma and Hepatic Lipids and Change Activities of Hepatic Antioxidant Enzymes in High Cholesterol-Fed Rats. International Journal for Vitamin and Nutrition Research. 2002, 72 (3),
  2. Keerthi Srinivas; Jerry W. King; Luke R. Howard; Jeana K. Monrad. Solubility and solution thermodynamic properties of quercetin and quercetin dihydrate in subcritical water. Journal of Food Engineering. 2010, 100 (2), 208-218