Thermo Scientific Chemicals

Methyl formate, 97%, may cont. up to ca 3% methanol, Thermo Scientific Chemicals

Catalog number: A15850.AP
500 mL, Each
Thermo Scientific Chemicals

Methyl formate, 97%, may cont. up to ca 3% methanol, Thermo Scientific Chemicals

Catalog number: A15850.AP
500 mL, Each
Quantity
Catalog number: A15850.AP
also known as A15850-AP
Price (USD)
Price: 32.70
Your price:
Quantity
-

Chemical Identifiers

CAS
107-31-3
IUPAC Name
methyl formate
Molecular Formula
C2H4O2
InChI Key
TZIHFWKZFHZASV-UHFFFAOYSA-N
SMILES
COC=O
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3415-1.3465 @ 20?C
Assay (GC)
≥96.0%

Description

Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text