4-Hydroxyquinoline, 98%
4-Hydroxyquinoline, 98%
4-Hydroxyquinoline, 98%
Thermo Scientific Chemicals

4-Hydroxyquinoline, 98%

CAS: 611-36-9 | C9H7NO | 145.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number A15859.06
also known as A15859-06
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS611-36-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or cream to pale brown
Melting Point (clear melt)202-206?C
Assay from Supplier's CofA≥97.5%
FormCrystals or needles or powder or crystalline powder
4-Hydroxyquinoline is a medical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyquinoline is a medical raw material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Charles R. Hauser.; George A. Reynolds. Reactions of β-Keto Esters with Aromatic Amines. Syntheses of 2- and 4-Hydroxyquinoline Derivatives. J. Am. Chem. Soc. 1948, 70 (7),2402-2404 .
  2. Jae Nyoung Kim.; Ka Young Lee.; Hyoung Shik Kim.; Tae Yi Kim. Synthesis of 3-Ethoxycarbonyl-4-hydroxyquinoline N-Oxides from the Baylis-Hillman Adducts of o-NitrobenzaldehydesOrg. Lett. 2000, 2 (3),343-345 .