2,3,4-Trichlorobenzenesulfonyl chloride, 95%
2,3,4-Trichlorobenzenesulfonyl chloride, 95%
2,3,4-Trichlorobenzenesulfonyl chloride, 95%
2,3,4-Trichlorobenzenesulfonyl chloride, 95%
Thermo Scientific Chemicals

2,3,4-Trichlorobenzenesulfonyl chloride, 95%

CAS: 34732-09-7 | C6H2Cl4O2S | 279.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A15865.03
also known as A15865-03
Price (USD)
35.65
Online Exclusive
39.40
Save 3.75 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.40
Save 3.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS34732-09-7
IUPAC Name2,3,4-trichlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H2Cl4O2S
InChI KeyJDAJYNHGBUXIKS-UHFFFAOYSA-N
SMILESClC1=CC=C(C(Cl)=C1Cl)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Assay (GC)≥94.0%
Melting Point (clear melt)57.0-66.0°C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only