Tetrahydro-4H-thiopyran-4-one, 99%
Tetrahydro-4H-thiopyran-4-one, 99%
Tetrahydro-4H-thiopyran-4-one, 99%
Thermo Scientific Chemicals

Tetrahydro-4H-thiopyran-4-one, 99%

CAS: 1072-72-6 | C5H8OS | 116.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A15872.03
also known as A15872-03
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.10
Save 6.45 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.10
Save 6.45 (15%)
Each
Chemical Identifiers
CAS1072-72-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)59.0-65.0?C
Assay (GC)≥98.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only