3,4-Dichlorobenzoic acid, 99%
3,4-Dichlorobenzoic acid, 99%
3,4-Dichlorobenzoic acid, 99%
3,4-Dichlorobenzoic acid, 99%
Thermo Scientific Chemicals

3,4-Dichlorobenzoic acid, 99%

CAS: 51-44-5 | C7H4Cl2O2 | 191.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15873.22
also known as A15873-22
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.20
Save 10.55 (14%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.20
Save 10.55 (14%)
Each
Add to cart
Chemical Identifiers
CAS51-44-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)203.5-209.5?C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only