I'm using the TaqMan hPSC Scorecard Panel. How do I set up my experimental run?
The template files contain all of the necessary experimental details for you to run your instrument. There is no need to modify any of the experimental details if you are using the template file (.edt). Step-by-step guides on how to run the qPCR instruments are available here.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center