1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
RUO – Research Use Only
General References:
- G.A. Crowder.; Mohammad-Reza Jalilian. Vibrational analysis of alkyl iodides: 1-iodo-2- methylpropane and 1-iodo-3-methylbutane. Journal of Molecular Structure. 1978, 49 (2), 287-292.
- J. R. Durig.; S. E. Godbey.; J. F. Sullivan. Far-infrared spectra, vibrational assignment, and conformational stability of 1-iodo-2-methylpropane. J. Phys. Chem. 2014, 92 (2), 6908-6913.