4-Bromo-2-fluorotoluene, 99%
4-Bromo-2-fluorotoluene, 99%
4-Bromo-2-fluorotoluene, 99%
Thermo Scientific Chemicals

4-Bromo-2-fluorotoluene, 99%

CAS: 51436-99-8 | C7H6BrF | 189.027 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15891.06
also known as A15891-06
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.40
Save 4.75
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.40
Save 4.75
Each
Chemical Identifiers
CAS51436-99-8
IUPAC Name4-bromo-2-fluoro-1-methylbenzene
Molecular FormulaC7H6BrF
InChI KeyYZFVUQSAJMLFOZ-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5270-1.5300 @ 20?C
FormLiquid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only