3-(Trifluoromethyl)aniline, 99%
3-(Trifluoromethyl)aniline, 99%
3-(Trifluoromethyl)aniline, 99%
3-(Trifluoromethyl)aniline, 99%
Thermo Scientific Chemicals

3-(Trifluoromethyl)aniline, 99%

CAS: 98-16-8 | C7H6F3N | 161.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A15910.36
also known as A15910-36
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS98-16-8
IUPAC Name3-(trifluoromethyl)aniline
Molecular FormulaC7H6F3N
InChI KeyVIUDTWATMPPKEL-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥98.5%
Refractive Index1.4785-1.4815 @ 20°C
FormLiquid
Identification (FTIR)Conforms
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. [5 g/L (20°C)]

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Domenico Spinelli .; Paolo Zanirato.; Elvira Di Miceli.; Liliana Lamartina.;Francesco Guerrera. On the Reaction of 3-Bromo-2-nitrobenzo[b]thiophene 13C-Labeled at C-2 with 3-(Trifluoromethyl)aniline: A Preliminary Insight into a Nucleophilic Substitution with Rearrangement. J. Org. Chem. 1997, 62 (15), 4921-4923 .
  2. V. Arjunan.; T. Rani.; S. Mohan. Spectroscopic and quantum chemical electronic structure investigations of 2-(trifluoromethyl)aniline and 3-(trifluoromethyl)aniline.J. Mol. Struct. 2011, (1-3), 179-193 .
  3. Used in an example of a general method for the conversion of aromatic amines to sulfonyl chlorides by reaction of the diazonium chlorides with SO2: Org. Synth. Coll., 7, 508 (1990).