1-Bromo-4-n-hexylbenzene, 97%
1-Bromo-4-n-hexylbenzene, 97%
1-Bromo-4-n-hexylbenzene, 97%
1-Bromo-4-n-hexylbenzene, 97%
Thermo Scientific Chemicals

1-Bromo-4-n-hexylbenzene, 97%

CAS: 23703-22-2 | C12H17Br | 241.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15918.06
also known as A15918-06
Price (USD)
89.65
Special offer
Online exclusive
Ends: 30-Jun-2026
105.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
89.65
Special offer
Online exclusive
Ends: 30-Jun-2026
105.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS23703-22-2
IUPAC Name1-bromo-4-hexylbenzene
Molecular FormulaC12H17Br
InChI KeyMXHOLIARBWJKCR-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.5185-1.5235 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only