Thermo Scientific Chemicals

1-Bromo-4-tert-butylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: A15924.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-4-tert-butylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: A15924.14
25 g, Each
Quantity
Catalog number: A15924.14
also known as A15924-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
3972-65-4
IUPAC Name
1-bromo-4-tert-butylbenzene
Molecular Formula
C10H13Br
InChI Key
XHCAGOVGSDHHNP-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(Br)C=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.5310-1.5350 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text