3,4-Dihydroxycinnamic acid, predominantly trans, 98+%
3,4-Dihydroxycinnamic acid, predominantly trans, 98+%
3,4-Dihydroxycinnamic acid, predominantly trans, 98+%
Thermo Scientific Chemicals

3,4-Dihydroxycinnamic acid, predominantly trans, 98+%

CAS: 331-39-5 | C9H8O4 | 180.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A15950.09
also known as A15950-09
Price (USD)
79.50
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
79.50
Each
Chemical Identifiers
CAS331-39-5
IUPAC Name(2Z)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Molecular FormulaC9H8O4
InChI KeyQAIPRVGONGVQAS-RQOWECAXSA-N
SMILESOC(=O)\C=C/C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.0%
Identification (FTIR)Conforms
Melting Pointca 195-210?C dec.
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White to cream or yellow or pale brown
View more
3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

Solubility
Soluble in DMSO (40 mg/mol), ethanol (25 mg/mL warm), DMF (∼7 mg/mL), PBS, PH 7.2 (∼0.65 mg/mol), hot water, ethyl acetate, Chloroform Hexane, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Frank Mercer.; Tim Goodman.; Janusz Wojtowicz.; David Duff. Synthesis and characterization of fluorinated aryl ethers prepared from decafluorobiphenyl. Journal of Polymer Science Part A: Polymer Chemistry. 1992, 30 (8), 1767-1770 .
  2. R Amadelli.; A De Battisti.; D.V Girenko.; S.V Kovalyov.; A.B Velichenko. Electrochemical oxidation of trans-3,4-dihydroxycinnamic acid at PbO2 electrodes: direct electrolysis and ozone mediated reactions compared. Electrochimica Acta. 2000, 46 (2-3), 341-347.