Thiomorpholine, 98%
Thiomorpholine, 98%
Thiomorpholine, 98%
Thermo Scientific Chemicals

Thiomorpholine, 98%

CAS: 123-90-0 | C4H9NS | 103.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15958.06
also known as A15958-06
Price (USD)
60.65
Online Exclusive
66.70
Save 6.05 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
66.70
Save 6.05 (9%)
Each
Chemical Identifiers
CAS123-90-0
IUPAC Namehydrogen thiomorpholine chloride
Molecular FormulaC4H10ClNS
InChI KeyQSJAHJYXDRUZMY-UHFFFAOYSA-N
SMILES[H+].[Cl-].C1CSCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
FormLiquid
Refractive Index1.5370-1.5410 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

Solubility
Miscible with waterMiscible with water and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.
RUO – Research Use Only

General References:

  1. Tooulia, K. K.; Theodosis-Nobelos, P.; Rekka, E. A. Thiomorpholine Derivatives with Hypolipidemic and Antioxidant Activity. Arch. Pharm. 2015, 348 (9), 629-634.
  2. Kacan, M.; Turkyilmaz, M.; Karabulut, F.; Altun, O.; Baran, Y. Complexation, thermal and catalytic studies of N-substituted piperazine, morpholine and thiomorpholine with some metal ions. Spectrochim. Acta, Part A 2014, 118, 572-577.