We offer the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit (Cat. Nos. A16517, A16518) which is a non-integrating system that uses Sendai virus vectors to reprogram somatic cells into induced pluripotent stem cells (iPSCs). The Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit contains three Invitrogen CytoTune 2.0 reprogramming vectors that are used for delivering and expressing key genetic factors necessary for reprogramming somatic cells into iPSCs. Only one application of the vectors is required for successful reprogramming.
Additionally, we offer the Episomal iPSC Reprogramming Vectors (Cat. No A14703) or Epi5 Episomal iPSC Reprogramming Kit (Cat. No A15960). This is a non-integrating system that reprograms somatic cells into induced pluripotent stem cells (iPSCs). These two products contain a mixture of six or five vectors designed to provide the optimal system for generating transgene-free and virus-free iPSCs in a feeder-free environment. Originally developed by Junying Yu and James Thomson (http://www.ncbi.nlm.nih.gov/pubmed/19325077) and further optimized by Cellular Dynamics International, these Episomal iPSC Reprogramming Vectors have proven successful in reprogramming a number of different somatic cell types.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.