2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol
2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol
2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol
Thermo Scientific Chemicals

2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol

CAS: 769-25-5 | C11H14 | 146.233 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15974.14
also known as A15974-14
Price (USD)
355.65
Special offer
Online exclusive
Ends: 30-Jun-2026
418.00
Save 62.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
355.65
Special offer
Online exclusive
Ends: 30-Jun-2026
418.00
Save 62.35 (15%)
Each
Chemical Identifiers
CAS769-25-5
IUPAC Name2-ethenyl-1,3,5-trimethylbenzene
Molecular FormulaC11H14
InChI KeyPDELBHCVXBSVPJ-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C=C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test5% in diethyl ether: clear colorless
Assay (GC)≥94.0%
FormLiquid
Refractive Index1.5265-1.5325 @ 20°C
Solution Test5% in methanol: clear colorless
View more
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

Solubility
Soluble in methanol.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Herbert Mayr; Armin R.Ofial; Holger Schimmel. Propagation rate of the cationic polymerization of 2,4,6-trimethylstyrene: A linear free energy approach. Macromolecules. 2005, 38, (1),33-40
  2. Cyrus O.Guss. The reaction of 2,4,6-trimethylstyrene oxide with phenol1. J. Am. Chem. Soc. 1953, 75, (13),3177-3179