2',4'-Dimethoxyacetophenone, 98%
2',4'-Dimethoxyacetophenone, 98%
2',4'-Dimethoxyacetophenone, 98%
Thermo Scientific Chemicals

2',4'-Dimethoxyacetophenone, 98%

CAS: 829-20-9 | C10H12O3 | 180.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A15982.22
also known as A15982-22
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS829-20-9
IUPAC Name1-(2,4-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O3
InChI KeyVQTDPCRSXHFMOL-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(C)=O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)36-42?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only