Mucobromic acid, 98+%
Mucobromic acid, 98+%
Mucobromic acid, 98+%
Mucobromic acid, 98+%
Thermo Scientific Chemicals

Mucobromic acid, 98+%

CAS: 488-11-9 | C4HBr2O3- | 256.857 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A15998.22
also known as A15998-22
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.30
Save 13.65 (14%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.30
Save 13.65 (14%)
Each
Add to cart
Chemical Identifiers
CAS488-11-9
IUPAC Name(2Z)-2,3-dibromo-4-oxobut-2-enoate
Molecular FormulaC4HBr2O3
InChI KeyNCNYEGJDGNOYJX-IHWYPQMZSA-M
SMILES[O-]C(=O)C(\Br)=C(\Br)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)119.0-126.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only