Benzo-15-crown-5, 98%
Benzo-15-crown-5, 98%
Benzo-15-crown-5, 98%
Benzo-15-crown-5, 98%
Thermo Scientific Chemicals

Benzo-15-crown-5, 98%

CAS: 14098-44-3 | C14H20O5 | 268.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16011.06
also known as A16011-06
Price (USD)
132.65
Special offer
Online exclusive
Ends: 30-Jun-2026
156.00
Save 23.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
132.65
Special offer
Online exclusive
Ends: 30-Jun-2026
156.00
Save 23.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS14098-44-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)75.0-83.0°C
Benzo-15-crown-5 is used in synthetic chemistry. It plays a major role and involved in complexation through ether oxygen. It is used in phase-transfer catalyst system. It is involved in 'Host-Guest' chemistry to identify the move of essential elements in the body. It can play the part of very complicated biological reactions such as enzyme functions, which finds application in the development of new pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo-15-crown-5 is used in synthetic chemistry. It plays a major role and involved in complexation through ether oxygen. It is used in phase-transfer catalyst system. It is involved in ′Host-Guest′ chemistry to identify the move of essential elements in the body. It can play the part of very complicated biological reactions such as enzyme functions, which finds application in the development of new pharmaceuticals.

Solubility
Soluble in hot methanol. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

General References:

  1. Inokuchi, Y.; Ebata, T.; Rizzo, T. R. Solvent Effects on the Encapsulation of Divalent Ions by Benzo-18-Crown-6 and Benzo-15-Crown-5. J. Phys. Chem. A 2015, 119 (29), 8097-8105.
  2. Hong, B.; Yang, F.; Ye, J.; Guo, H.; Yan, X. Novel Benzo-15-crown-5/C60 Dyads with Different Chains: Synthesis and Complexation Properties. Lett. Org. Chem. 2014, 11 (1), 2-5.