Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP - FAQs
Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP - FAQs
View additional product information for Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP - FAQs (A16020)
1 product FAQs found
I used the RiboMinus Eukaryote System v2 for total transcriptome isolation but I am seeing genomic DNA contamination. Why is this?
This is most likely due to contamination of the total RNA sample with genomic DNA. Before the RiboMinus Kit procedure, we recommend treating the total RNA sample with DNase I to remove any genomic DNA contamination.