N(^e)-Benzyloxycarbonyl-L-lysine, 98%
N(^e)-Benzyloxycarbonyl-L-lysine, 98%
N(^e)-Benzyloxycarbonyl-L-lysine, 98%
N(^e)-Benzyloxycarbonyl-L-lysine, 98%
Thermo Scientific Chemicals

N(^e)-Benzyloxycarbonyl-L-lysine, 98%

CAS: 1155-64-2 | C14H20N2O4 | 280.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16022.06
also known as A16022-06
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.10
Save 6.45 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.10
Save 6.45 (15%)
Each
Add to cart
Chemical Identifiers
CAS1155-64-2
IUPAC Name(2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular FormulaC14H20N2O4
InChI KeyCKGCFBNYQJDIGS-LBPRGKRZSA-N
SMILESN[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
Optical Rotation15.3 ± 0.5? (C=1 in 1N HCl)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only