trans-3-Bromocinnamic acid, 98+%
trans-3-Bromocinnamic acid, 98+%
trans-3-Bromocinnamic acid, 98+%
trans-3-Bromocinnamic acid, 98+%
Thermo Scientific Chemicals

trans-3-Bromocinnamic acid, 98+%

CAS: 14473-91-7 | C9H7BrO2 | 227.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16035.14
also known as A16035-14
Price (USD)
147.65
Online Exclusive
164.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
147.65
Online Exclusive
164.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS14473-91-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
FormPowder
Melting Point (clear melt)176.0-182.0?C
trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Takayoshi Suzuki.; Keiko Imai.; Erika Imai.; Shinsuke Iida.; Ryuzo Ueda.; Hiroki Tsumoto.; Hidehiko Nakagawa.; Naoki Miyata. Design, synthesis, enzyme inhibition, and tumor cell growth inhibition of 2-anilinobenzamide derivatives as SIRT1 inhibitors. Progress in Organic Coatings. 2009, 17 (16), 5900-5905.