Bis(2-ethylhexyl) sebacate, 95%
Bis(2-ethylhexyl) sebacate, 95%
Bis(2-ethylhexyl) sebacate, 95%
Thermo Scientific Chemicals

Bis(2-ethylhexyl) sebacate, 95%

CAS: 122-62-3 | C26H50O4 | 426.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
5000 mL
Catalog number A16049.0J
also known as A16049-0J
Price (USD)
257.65
Special offer
Online exclusive
Ends: 30-Jun-2026
303.00
Save 45.35 (15%)
Each
Quantity:
5000 mL
Request bulk or custom format
Price (USD)
257.65
Special offer
Online exclusive
Ends: 30-Jun-2026
303.00
Save 45.35 (15%)
Each
Chemical Identifiers
CAS122-62-3
IUPAC Name1,10-bis(2-ethylhexyl) decanedioate
Molecular FormulaC26H50O4
InChI KeyVJHINFRRDQUWOJ-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
Refractive Index1.4475-1.4535 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Markus Rothmaier, Wilhelm Simon. Chloride-selective electrodes based on mercury organic compounds as neutral carriers. Analytica Chimica Acta. 1993, 271 (1), 135-141.
  2. Yuji Kawabata.; Toshito Kamichika.; Totaro Imasaka.; Nobuhiko Ishibashi. Ion-selective optrode using hexadecyl-Acridine Orange attached on poly(vinyl chloride) membrane. Anal. Chem. 1990, 62 (18), 2054-2055.