Methylcyclohexane, 99%
Methylcyclohexane, 99%
Methylcyclohexane, 99%
Methylcyclohexane, 99%
Thermo Scientific™

Methylcyclohexane, 99%

CAS: 108-87-2 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number A16057.AP
also known as A16057-AP
Price (USD)
79.10
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
79.10
Each
Add to cart
Chemical Identifiers
CAS108-87-2
IUPAC Namemethylcyclohexane
Molecular FormulaC7H14
InChI KeyUAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILESCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4215-1.4245 @ 20°C (non-U.S. specification)
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms (non-U.S. specification)
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. For use as a solvent in the preparation of unsolvated Grignard reagents from an alkyl halide and Mg powder: Org. Synth. Coll., 5, 1141 (1973).
  2. Narayanaswamy, K.; Pitsch, H.; Pepiot, P. A chemical mechanism for low to high temperature oxidation of methylcyclohexane as a component of transportation fuel surrogates. Combust. Flame 2015, 162 (4), 1193-1213.
  3. Ngoye, F.; Lakiss, L.; Qin, Z.; Laforge, S.; Canaff, C.; Tarighi, M.; Valtchev, V.; Thomas, K.; Vicente, A.; Gilson, J. Mitigating coking during methylcyclohexane transformation on HZSM-5 zeolites with additional porosity. J. Catal. 2014, 320, 118-126.