Thermo Scientific Chemicals

Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals

Catalog number: A16068.0J
5000 mL, Each
Thermo Scientific Chemicals

Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals

Catalog number: A16068.0J
5000 mL, Each
Quantity
Catalog number: A16068.0J
also known as A16068-0J
Price (USD)
Price: 121.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
112-34-5
IUPAC Name
2-(2-butoxyethoxy)ethan-1-ol
Molecular Formula
C8H18O3
InChI Key
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
SMILES
CCCCOCCOCCO
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤0.5%
Refractive Index
1.4305-1.4335 @ 20?C

Description

Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.

Solubility
Miscible with water, ethyl ether, ethanol acetone and benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents and light metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text