Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP - FAQs
Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP - FAQs
View additional product information for Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP - FAQs (A16110SAMPLE, A16110)
1 product FAQs found
Can you provide the shelf life for Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP (Cat. No. A16110)?
Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP (Cat. No. A16110) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.