4-Bromo-2-fluorobenzaldehyde, 97%
4-Bromo-2-fluorobenzaldehyde, 97%
4-Bromo-2-fluorobenzaldehyde, 97%
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzaldehyde, 97%

CAS: 57848-46-1 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16112.14
also known as A16112-14
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS57848-46-1
IUPAC Name4-bromo-2-fluorobenzaldehyde
Molecular FormulaC7H4BrFO
InChI KeyUPCARQPLANFGQJ-UHFFFAOYSA-N
SMILESFC1=CC(Br)=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystalline powder or powder
Assay (GC)≥96.0%
Free acid (titration)≤1.5%
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.

Solubility
Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases. Keep the container tightly closed.
RUO – Research Use Only

General References:

  1. Jagadhani, S. G.; Kundlikar, S. G.; Karale, B. K. Synthesis and Characterization of Some Fluorinated 1,5-Benzothiazepines and Pyrazolines. Orient. J. Chem. 2015, 31 (1), 601-604.
  2. Trost, B. M.; Osipov, M.; Krüger, S.; Zhang, Y. A catalytic asymmetric total synthesis of (-)-perophoramidine. Chem. Sci. 2015, 6, 349-353.