alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
Thermo Scientific Chemicals

alpha-Naphtholbenzein

CAS: 145-50-6 | C27H18O2 | 374.439 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16125.22
also known as A16125-22
Price (USD)
418.65
Online Exclusive
465.00
Save 46.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
418.65
Online Exclusive
465.00
Save 46.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS145-50-6
IUPAC Name(4E)-4-[(4-hydroxynaphthalen-1-yl)(phenyl)methylidene]-1,4-dihydronaphthalen-1-one
Molecular FormulaC27H18O2
InChI KeyVDDWRTZCUJCDJM-SOYKGTTHSA-N
SMILESOC1=C2C=CC=CC2=C(C=C1)C(\C1=CC=CC=C1)=C1/C=CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Color ChangeOrange @ pH8.2 to blue-green @ pH10.0
Specific Absorbance (E 1%/1cm)≥350
CommentSuitable for use as an indicator
lambda-max (UV-Vis)644-652nm (0.01M NaOH)
View more
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.
RUO – Research Use Only

General References:

  1. Acid-base indicator: pH 0.0 - 0.8; 8.2 - 10.0.
  2. Wang, J.; Xu, C.; Song, Q. Rapid and Environmentally Friendly Three-Dimensional-Printed Flow Injection Analysis System for the Determination of the Acid Number in Thermal Conductive Oil. Energy Fuels 2015, 29 (2), 1040-1044.
  3. Özçeşmeci, M. Synthesis, photophysical and photochemical properties of metal-free and zinc(II) phthalocyanines bearing alpha-naphtholbenzein units. J. Organomet. Chem. 2014, 767, 16-21.