Glyoxal, 40% w/w aq. soln.
Glyoxal, 40% w/w aq. soln.
Glyoxal, 40% w/w aq. soln.
Thermo Scientific Chemicals

Glyoxal, 40% w/w aq. soln.

CAS: 107-22-2 | C2H2O2 | 58.036 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
100 mL
500 mL
2500 mL
Catalog number A16144.0D
Price (USD)
357.65
Special offer
Online exclusive
Ends: 28-Dec-2026
421.00
Save 63.35
Each
Quantity:
10,000 mL
Request bulk or custom format
Price (USD)
357.65
Special offer
Online exclusive
Ends: 28-Dec-2026
421.00
Save 63.35
Each
Chemical Identifiers
CAS107-22-2
IUPAC Nameoxaldehyde
Molecular FormulaC2H2O2
InChI KeyLEQAOMBKQFMDFZ-UHFFFAOYSA-N
SMILESO=CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (unspecified)≥39.5 to ≤40.5%
Identification (FTIR)Conforms
Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.

Solubility
Miscible with water, methanol and dimethylsulfoxide.

Notes
Moisture and heat sensitive. Incompatible with strong oxidizing agents, strong reducing agent, oxygen, peroxides, amides, amines, hydroxy-containing materials, nitric acid and aldehydes.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. For use in the chemoselective hydrolysis of amides in the presence of esters, see Copper(II) chloride dihydrate, A16064 .
  2. For use in the synthesis of bicyclic diketones, see Dimethyl acetone-1,3-dicarboxyl ate, A14969 .
  3. Kurten, T.; Elm, J.; Prisle, N. L.; Mikkelsen, K. V.; Kampf, C. J.; Waxman, E. M.; Volkamer, R. Computational Study of the Effect of Glyoxal-Sulfate Clustering on the Henry’s Law Coefficient of Glyoxal. J. Phys. Chem. A 2015, 119 (19), 4509-4514.
  4. Lawson, S. J.; Selleck, P. W.; Galbally, I. E.; Keywood, M. D.; Harvey, M. J.; Lerot, C.; Helmig, D.; Ristovski, Z. Seasonal in situ observations of glyoxal and methylglyoxal over the temperate oceans of the Southern Hemisphere. Atmos. Chem. Phys. 2015, 15 (1), 223-240.