Lithium metaborate, dried, 99%
Lithium metaborate, dried, 99%
Lithium metaborate, dried, 99%
Lithium metaborate, dried, 99%
Thermo Scientific Chemicals

Lithium metaborate, dried, 99%

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16145.36
also known as A16145-36
Price (USD)
341.65
Special offer
Online exclusive
Ends: 30-Jun-2026
402.00
Save 60.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
341.65
Special offer
Online exclusive
Ends: 30-Jun-2026
402.00
Save 60.35 (15%)
Each
Chemical Identifiers
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)>98.5% to <101.5%
Loss on Drying<2.5% (200?C, constant. wt)
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

Solubility
Soluble in water(27.5g/L).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alkali metals, acid anhydrides, acids.
RUO – Research Use Only

General References:

  1. Gwendy E.M. Hall,; J-C. Pelchat. Analysis of Standard Reference Materials for Zr, Nb, Hf and Ta by ICP-MS after Lithium Metaborate Fusion and Cupferron Separation*. Journal name. 1990, 41 (1), 197-206.
  2. A.A. Verbeek,; M.C. Mitchell,; A.M. Ure. The analysis of small samples of rock and soil by atomic absorption and emission spectrometry after a lithium metaborate fusionitric acid dissolution procedure . Analytica Chimica Acta. 1982, 135 (2) ,215-228.