Benzenesulfonic acid, 94%
Benzenesulfonic acid, 94%
Benzenesulfonic acid, 94%
Thermo Scientific Chemicals

Benzenesulfonic acid, 94%

CAS: 98-11-3 | C6H6O3S | 158.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16148.0E
also known as A16148-0E
Price (USD)
95.60
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
95.60
Each
Chemical Identifiers
CAS98-11-3
IUPAC Namebenzenesulfonic acid
Molecular FormulaC6H6O3S
InChI KeySRSXLGNVWSONIS-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)> 92.5%
Water Content (Karl Fischer Titration)< 3.0%
Assay (Silylated GC)> 92.5%
Appearance (Color)White to brown
Formsolid
Benzenesulfonic acid is mainly consumed by conversion to other specialty chemicals. A variety of pharmaceutical drugs are prepared as salts of benzenesulfonic acid and are known as besylates or besilates. The alkali metal salt of benzenesulfonic acid was once widely used in the production of phenol. It acts as an acid catalyst for direct esterification of amino acids and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzenesulfonic acid is mainly consumed by conversion to other specialty chemicals. A variety of pharmaceutical drugs are prepared as salts of benzenesulfonic acid and are known as besylates or besilates. The alkali metal salt of benzenesulfonic acid was once widely used in the production of phenol. It acts as an acid catalyst for direct esterification of amino acids and peptides.

Solubility
Soluble in water and alcohol. Slightly soluble in benzene. Insoluble in non-polar solvents.

Notes
It is hygroscopic. Incompatible with oxidizing agents and bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Sanjay P.Kamble; Sudhir B.Sawant; Vishwas G.Pangarkar . Batch and continuous photocatalytic degradation of benzenesulfonic acid using concentrated solar radiation. Ind. Eng. Chem. Res. 2003, 42, (26),6705-6713
  2. Hans Cerfontain; Ankie Koeberg-Telder; Cornelis Kruk. Solutes in sulfuric acid. Part VII. Ionization of benzenesulfonic acid; determination of pKBH by 13C NMR. Tetrahedron Letters. 1975, 16, (42),3639-3642