Strontium acetate hemihydrate, 98%
Strontium acetate hemihydrate, 98%
Strontium acetate hemihydrate, 98%
Thermo Scientific Chemicals

Strontium acetate hemihydrate, 98%

CAS: 14692-29-6 | Sr(OOCCH3)2·0.5H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16149.36
also known as A16149-36
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS14692-29-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Y. Duan,J. Li, X. Yang, X. -M. Cao, L. Hu, Z. -Y. Wang, Y. -W. Liu , C. -X. Wang.Thermal investigation of strontium acetate hemihydrate in nitrogen gas . Journal of Thermal Analysis and Calorimetry. 2008, 941, 169-174.
  2. Sushil K. Misra,; Krishan Kumar. Single crystal EPR study of Cu2+-doped strontium acetate hemihydrate [Sr(CH3COO)2 · Full-size image (<1 K)H2O] . Physica B+C. 1986, 138 (1-2), 171-175.