Potassium iodate, 98%
Potassium iodate, 98%
Potassium iodate, 98%
Thermo Scientific Chemicals

Potassium iodate, 98%

Potassium iodate, 98%, IKO3, CAS Number-7758-05-6, iodic acid, potassium salt, kaliumjodat, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, potassium iodate, iodic acid hio3 , potassium salt, caswell no. 693a, potassium triodate, potassium iodine oxide kio3, unii-i139e44nhl | CAS: 7758-05-6 | IKO3 | 214.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16162.36
also known as A16162-36
Price (USD)
174.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
174.00
Each
Chemical Identifiers
CAS7758-05-6
IUPAC Namepotassium iodate
Molecular FormulaIKO3
InChI KeyJLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)≥97.5 to ≤102.5%
Appearance (Color)White
Loss on Drying<1.0% (200?C, constant weight)
Identification (FTIR)Conforms
FormPowder
Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

Solubility
Soluble in water, potassium iodide solution. Insoluble in alcohol, liquid ammonia and nitric acid.

Notes
Incompatible with aluminum, strong oxidizing agents, sulfides, peroxides, metals and reducing agents. Avoid heat, shock and friction.
RUO – Research Use Only

General References:

  1. Tang, Y.; Li, Y.; Yu, Z.; Bai, Y.; Chen, Y.; Sun, Y.; Wan, P. Energy-saving synthesis of potassium iodate via electrolysis of potassium iodine and O2 in a membraneless cell. Green Chem. 2012, 14 (2), 334-337.
  2. Muraleedharan, K. Thermal decomposition kinetics of potassium iodate. J. Therm. Anal. Calorim. 2011, 103 (3), 943-955.